Product Details
- SNP ID
-
rs3668074
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:54986423 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCTGGTACCTAAGGATCTGCGC[C/A]CACCCTTCCGTTACATGAAAGGCCG
- Phenotype
-
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Sf3b1
PubMed Links
Gene Details
- Gene
- Sf3b1
- Gene Name
- splicing factor 3b, subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031179.2 |
4938 |
UTR 3 |
|
|
NP_112456.2 |
View Full Product Details