Product Details
- SNP ID
-
rs4222507
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:89153267 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACCAGGCGCTGGAAGCACCTCAC[A/G]GGGACACTGATCCTGGTGAACTCAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sh3bp4
PubMed Links
Gene Details
- Gene
- Sh3bp4
- Gene Name
- SH3-domain binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_133816.2 |
3147 |
Silent Mutation |
|
|
NP_598577.1 |
View Full Product Details