Product Details
- SNP ID
-
rs3685325
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:8451874 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCCGACATGGTATGTAACTCTC[C/G]GGTTACTTAGAAACGGTCCCAAAGG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
Tns3
PubMed Links
Gene Details
- Gene
- Tns3
- Gene Name
- tensin 3
View Full Product Details