Product Details

SNP ID
rs3023422
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:79484266 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCAGGGCGGAGTACCTGGGTCCTT[C/T]CCAGTCCTGGGCCCTGTAGTCAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Kcnj4 PubMed Links

Gene Details

Gene
Kcnj4
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008427.4 1604 UTR 3 NP_032453.3

View Full Product Details