Product Details

SNP ID
rs4225888
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:49128511 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAGACAGGAAGATAGAGAGGA[A/G]CCAGAGCCCTAAGCCTCTCTGTACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Igf2bp3 PubMed Links

Gene Details

Gene
Igf2bp3
Gene Name
insulin-like growth factor 2 mRNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023670.3 Intron NP_076159.3
Gene
Malsu1
Gene Name
mitochondrial assembly of ribosomal large subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029353.1 Intron NP_083629.1

View Full Product Details