Assay Design



Gene Transcripts

Gene Symbol
AQP10
Entrez Gene ID
102118895
Gene Name
aquaporin 10
Gene Aliases
-
Chromosome Location
Chr.1: 97503881 - 97507762 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541671.2 XP_005541728.1 2 - 3 570 66

View Full Product Details