Assay Design



Gene Transcripts

Gene Symbol
MBLAC2
Entrez Gene ID
102142761
Gene Name
metallo-beta-lactamase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 88860091 - 88879244 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557359.2 XP_005557416.1 1 - 2 965 59

View Full Product Details