Assay Design



Gene Transcripts

Gene Symbol
KLHL23
Entrez Gene ID
102142348
Gene Name
kelch like family member 23
Gene Aliases
-
Chromosome Location
Chr.12: 58613036 - 58631069 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573424.2 XP_005573481.1 2 - 3 1477 140

View Full Product Details