Assay Design



Gene Transcripts

Gene Symbol
SGCZ
Entrez Gene ID
102132578
Gene Name
sarcoglycan zeta
Gene Aliases
-
Chromosome Location
Chr.8: 13993944 - 15195613 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562643.2 XP_005562700.1 2 - 3 1501 110

View Full Product Details