Assay Design



Gene Transcripts

Gene Symbol
CEACAM16
Entrez Gene ID
102134905
Gene Name
CEA cell adhesion molecule 16, tectorial membrane component
Gene Aliases
-
Chromosome Location
Chr.19: 45433546 - 45444738 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589544.2 XP_005589601.1 5 - 6 1142 72
XM_015441076.1 XP_015296562.1 4 - 5 990 72

View Full Product Details