Assay Design



Gene Transcripts

Gene Symbol
RAB33A
Entrez Gene ID
102119481
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.X: 127052076 - 127066791 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594560.2 XP_005594617.1 1 - 2 1106 69

View Full Product Details