Assay Design



Gene Transcripts

Gene Symbol
SYNDIG1L
Entrez Gene ID
102126812
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
-
Chromosome Location
Chr.7: 139095276 - 139115359 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561767.1 XP_005561824.1 3 - 4 786 70

View Full Product Details