Assay Design



Gene Transcripts

Gene Symbol
VSTM2L
Entrez Gene ID
102121226
Gene Name
V-set and transmembrane domain containing 2 like
Gene Aliases
-
Chromosome Location
Chr.10: 69483913 - 69524993 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568972.1 XP_005569029.1 1 - 2 371 70

View Full Product Details