Assay Design



Gene Transcripts

Gene Symbol
KCNC3
Entrez Gene ID
102115741
Gene Name
potassium voltage-gated channel subfamily C member 3
Gene Aliases
-
Chromosome Location
Chr.19: 51130165 - 51151571 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015441044.1 XP_015296530.1 3 - 4 1468 61

View Full Product Details