Assay Design



Gene Transcripts

Gene Symbol
SP9
Entrez Gene ID
102147150
Gene Name
Sp9 transcription factor
Gene Aliases
-
Chromosome Location
Chr.12: 63274536 - 63280140 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573516.2 XP_005573573.1 1 - 2 110 105

View Full Product Details