Assay Design



Gene Transcripts

Gene Symbol
GPHB5
Entrez Gene ID
102138045
Gene Name
glycoprotein hormone subunit beta 5
Gene Aliases
-
Chromosome Location
Chr.7: 127635881 - 127646609 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561445.2 XP_005561502.1 1 - 2 4079 135

View Full Product Details