Assay Design



Gene Transcripts

Gene Symbol
AIDA
Entrez Gene ID
101925904
Gene Name
axin interactor, dorsalization associated
Gene Aliases
-
Chromosome Location
Chr.1: 81508817 - 81551817 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283182.1 NP_001270111.1 1 - 2 682 88
XM_015440188.1 XP_015295674.1 2 - 3 392 88
GenBank mRNA AB168527.1 - 1 - 2 682 88

View Full Product Details