Assay Design



Gene Transcripts

Gene Symbol
BCL2
Entrez Gene ID
102138212
Gene Name
BCL2 apoptosis regulator
Gene Aliases
-
Chromosome Location
Chr.18: 17013626 - 17218351 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586525.1 XP_005586582.1 1 - 2 635 145

View Full Product Details