Assay Design



Gene Transcripts

Gene Symbol
CCL11
Entrez Gene ID
102136803
Gene Name
C-C motif chemokine ligand 11
Gene Aliases
-
Chromosome Location
Chr.16: 30646140 - 30648596 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583422.2 XP_005583479.1 2 - 3 332 71

View Full Product Details