Assay Design



Gene Transcripts

Gene Symbol
KLRF1
Entrez Gene ID
102141941
Gene Name
killer cell lectin like receptor F1
Gene Aliases
nkp80
Chromosome Location
Chr.11: 10175766 - 10191760 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285324.1 NP_001272253.1 3 - 4 347 69
XM_015430177.1 XP_015285663.1 2 - 3 869 69
GenBank mRNA AJ426430.1 - 3 - 4 347 69

View Full Product Details