Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005578188.2 | XP_005578245.1 | 7 - 8 | 1689 | 78 |
XM_005578189.2 | XP_005578246.1 | 7 - 8 | 1917 | 78 | |
XM_015434917.1 | XP_015290403.1 | 7 - 8 | 1159 | 78 | |
XM_015434918.1 | XP_015290404.1 | 7 - 8 | 1091 | 78 |