Assay Design



Gene Transcripts

Gene Symbol
TAX1BP3
Entrez Gene ID
102144212
Gene Name
Tax1 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.16: 3587937 - 3593088 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285076.1 NP_001272005.1 2 - 3 238 76
XM_015437388.1 XP_015292874.1 2 - 3 310 76
GenBank mRNA AB172355.1 - 2 - 3 238 76

View Full Product Details