Assay Design



Gene Transcripts

Gene Symbol
MT3
Entrez Gene ID
102134329
Gene Name
metallothionein 3
Gene Aliases
MT-3, MT-III
Chromosome Location
Chr.20: 44572346 - 44574195 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591968.2 XP_005592025.1 1 - 2 380 93
GenBank mRNA AB220447.2 - 1 - 2 97 93

View Full Product Details