Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005543591.2 | XP_005543648.1 | 2 - 3 | 348 | 84 |
XM_005543592.2 | XP_005543649.1 | 2 - 3 | 567 | 84 | |
XM_015436911.1 | XP_015292397.1 | 2 - 3 | 1009 | 84 | |
XM_015436916.1 | XP_015292402.1 | 2 - 3 | 276 | 84 | |
XM_015436918.1 | XP_015292404.1 | 3 - 4 | 770 | 84 |