Assay Design



Gene Transcripts

Gene Symbol
PTAR1
Entrez Gene ID
102125906
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 80449609 - 80500017 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581890.2 XP_005581947.1 1 - 2 157 57
XM_005581891.2 XP_005581948.1 1 - 2 160 57
XM_005581892.2 XP_005581949.1 1 - 2 160 57
XM_015437259.1 XP_015292745.1 1 - 2 160 57
XM_015437261.1 XP_015292747.1 1 - 2 179 57

View Full Product Details