Assay Design



Gene Transcripts

Gene Symbol
THAP9
Entrez Gene ID
102127636
Gene Name
THAP domain containing 9
Gene Aliases
-
Chromosome Location
Chr.5: 51505575 - 51525876 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554918.2 XP_005554975.1 2 - 3 432 122

View Full Product Details