Assay Design



Gene Transcripts

Gene Symbol
TMEM60
Entrez Gene ID
102146023
Gene Name
transmembrane protein 60
Gene Aliases
-
Chromosome Location
Chr.3: 134460878 - 134465763 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550389.2 XP_005550446.1 1 - 2 419 97

View Full Product Details