Assay Design



Gene Transcripts

Gene Symbol
HACD4
Entrez Gene ID
102117284
Gene Name
3-hydroxyacyl-CoA dehydratase 4
Gene Aliases
PTPLAD2
Chromosome Location
Chr.15: 57712279 - 57740701 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581637.2 XP_005581694.1 5 - 6 891 115

View Full Product Details