Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005542638.2 | XP_005542695.1 | 5 - 6 | 587 | 77 |
XM_005542642.2 | XP_005542699.1 | 6 - 7 | 698 | 77 | |
XM_005542644.2 | XP_005542701.1 | 5 - 6 | 737 | 77 | |
XM_005542646.2 | XP_005542703.1 | 5 - 6 | 570 | 77 | |
XM_005542647.2 | XP_005542704.1 | 5 - 6 | 552 | 77 | |
XM_005542649.2 | XP_005542706.1 | 5 - 6 | 1073 | 77 | |
XM_015432486.1 | XP_015287972.1 | 7 - 8 | 4042 | 77 | |
XM_015432488.1 | XP_015287974.1 | 7 - 8 | 3803 | 77 | |
XM_015432493.1 | XP_015287979.1 | 7 - 8 | 1145 | 77 | |
XM_015432498.1 | XP_015287984.1 | 5 - 6 | 765 | 77 | |
XM_015432509.1 | XP_015287995.1 | 6 - 7 | 609 | 77 |