Assay Design



Gene Transcripts

Gene Symbol
ALKBH2
Entrez Gene ID
102116761
Gene Name
alkB homolog 2, alpha-ketoglutarate dependent dioxygenase
Gene Aliases
-
Chromosome Location
Chr.11: 112731005 - 112736764 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572173.2 XP_005572230.1 2 - 3 811 90
XM_005572174.2 XP_005572231.1 2 - 3 553 90
XM_005572175.2 XP_005572232.1 2 - 3 563 90

View Full Product Details