Assay Design



Gene Transcripts

Gene Symbol
RPL23
Entrez Gene ID
102126462
Gene Name
ribosomal protein L23
Gene Aliases
-
Chromosome Location
Chr.16: 50355703 - 50359415 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583988.2 XP_005584045.2 4 - 5 473 128

View Full Product Details