Assay Design



Gene Transcripts

Gene Symbol
EPDR1
Entrez Gene ID
102132496
Gene Name
ependymin related 1
Gene Aliases
MERP-1, MERP1, QmoA-12340, QmoA-13475, QtrA-11871
Chromosome Location
Chr.3: 82532404 - 82566002 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549761.2 XP_005549818.1 1 - 2 631 93
GenBank mRNA AB046003.1 - 1 - 2 427 93
AB063094.1 - 1 - 2 555 93
AB066535.1 - 2 - 3 528 93
AB066537.1 - 1 - 2 446 93
AB097520.1 - 1 - 2 562 93

View Full Product Details