Assay Design



Gene Transcripts

Gene Symbol
ANP32B
Entrez Gene ID
102121176
Gene Name
acidic nuclear phosphoprotein 32 family member B
Gene Aliases
-
Chromosome Location
Chr.15: 39763558 - 39795661 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581222.2 XP_005581279.1 1 - 1 180 135
XM_015436956.1 XP_015292442.1 1 - 1 180 135

View Full Product Details