Assay Design



Gene Transcripts

Gene Symbol
RAB38
Entrez Gene ID
102136247
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.14: 83567358 - 83629956 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579305.2 XP_005579362.1 2 - 3 579 91

View Full Product Details