Assay Design



Gene Transcripts

Gene Symbol
LRRTM4
Entrez Gene ID
102146066
Gene Name
leucine rich repeat transmembrane neuronal 4
Gene Aliases
-
Chromosome Location
Chr.13: 32227911 - 33044384 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015433497.1 XP_015288983.1 2 - 3 460 100
XM_015433498.1 XP_015288984.1 2 - 3 460 100

View Full Product Details