Assay Design



Gene Transcripts

Gene Symbol
SOST
Entrez Gene ID
102130808
Gene Name
sclerostin
Gene Aliases
-
Chromosome Location
Chr.16: 55448006 - 55453104 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584371.2 XP_005584428.1 1 - 2 276 72

View Full Product Details