Assay Design



Gene Transcripts

Gene Symbol
GJA10
Entrez Gene ID
102146778
Gene Name
gap junction protein alpha 10
Gene Aliases
-
Chromosome Location
Chr.4: 84052112 - 84053743 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552352.1 XP_005552409.1 1 - 1 1457 120

View Full Product Details