Assay Design



Gene Transcripts

Gene Symbol
ARL11
Entrez Gene ID
102128648
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
-
Chromosome Location
Chr.17: 28830981 - 28836297 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585864.2 XP_005585921.1 1 - 2 543 160

View Full Product Details