Assay Design



Gene Transcripts

Gene Symbol
PRPS1L1
Entrez Gene ID
102124428
Gene Name
phosphoribosyl pyrophosphate synthetase 1 like 1
Gene Aliases
-
Chromosome Location
Chr.3: 102736068 - 102737144 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550040.2 XP_005550097.1 1 - 1 61 101

View Full Product Details