Assay Design



Gene Transcripts

Gene Symbol
LRRC66
Entrez Gene ID
102121373
Gene Name
leucine rich repeat containing 66
Gene Aliases
-
Chromosome Location
Chr.5: 82497842 - 82529446 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555306.2 XP_005555363.2 4 - 5 1907 141

View Full Product Details