Assay Design



Gene Transcripts

Gene Symbol
RGS21
Entrez Gene ID
102116803
Gene Name
regulator of G protein signaling 21
Gene Aliases
-
Chromosome Location
Chr.1: 50493766 - 50544678 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005540263.2 XP_005540320.1 1 - 2 162 77

View Full Product Details