Assay Design



Gene Transcripts

Gene Symbol
FGF17
Entrez Gene ID
102137925
Gene Name
fibroblast growth factor 17
Gene Aliases
-
Chromosome Location
Chr.8: 22174965 - 22181034 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562738.2 XP_005562795.1 1 - 2 215 85

View Full Product Details