Assay Design



Gene Transcripts

Gene Symbol
IL36RN
Entrez Gene ID
102124609
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
-
Chromosome Location
Chr.13: 17208189 - 17213572 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575302.2 XP_005575359.1 2 - 3 168 87
XM_015433400.1 XP_015288886.1 2 - 3 168 87

View Full Product Details