Assay Design



Gene Transcripts

Gene Symbol
TIMM22
Entrez Gene ID
102139290
Gene Name
translocase of inner mitochondrial membrane 22
Gene Aliases
-
Chromosome Location
Chr.16: 829787 - 836864 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582422.2 XP_005582479.1 2 - 3 1121 122

View Full Product Details