Assay Design



Gene Transcripts

Gene Symbol
ABT1
Entrez Gene ID
102145414
Gene Name
activator of basal transcription 1
Gene Aliases
-
Chromosome Location
Chr.4: 143966057 - 143968267 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553801.2 XP_005553858.1 2 - 3 478 64

View Full Product Details