Assay Design



Gene Transcripts

Gene Symbol
TMEM97
Entrez Gene ID
101865327
Gene Name
transmembrane protein 97
Gene Aliases
-
Chromosome Location
Chr.16: 24540136 - 24549588 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283922.1 NP_001270851.1 2 - 3 318 66
XM_015437797.1 XP_015293283.1 2 - 3 418 66
XM_015437798.1 XP_015293284.1 2 - 3 467 66
GenBank mRNA AB168549.1 - 2 - 3 318 66
AB170603.1 - 2 - 3 327 66

View Full Product Details