Assay Design



Gene Transcripts

Gene Symbol
N4BP3
Entrez Gene ID
102143557
Gene Name
NEDD4 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 177949443 - 177961527 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558689.2 XP_005558746.1 2 - 3 444 87

View Full Product Details