Assay Design



Gene Transcripts

Gene Symbol
UGT2A3
Entrez Gene ID
102124844
Gene Name
UDP glucuronosyltransferase family 2 member A3
Gene Aliases
-
Chromosome Location
Chr.5: 65277930 - 65309472 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555165.2 XP_005555222.1 2 - 3 893 158

View Full Product Details