Assay Design



Gene Transcripts

Gene Symbol
TUBAL3
Entrez Gene ID
102145991
Gene Name
tubulin alpha like 3
Gene Aliases
-
Chromosome Location
Chr.9: 5759713 - 5771571 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564551.2 XP_005564608.1 1 - 2 240 73

View Full Product Details