Assay Design



Gene Transcripts

Gene Symbol
LDHAL6A
Entrez Gene ID
102134594
Gene Name
lactate dehydrogenase A like 6A
Gene Aliases
-
Chromosome Location
Chr.14: 49190039 - 49216912 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578441.2 XP_005578498.1 7 - 8 1137 85

View Full Product Details